Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATTTGGGTATATAAGGCATTTG[C/T]AGTCAAAGAAAATAATTCACATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608110 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BUB1B-PAK6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BUB1B-PAK6 - BUB1B-PAK6 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128628.2 | 2039 | Intron | NP_001122100.1 | |||
NM_001128629.2 | 2039 | Intron | NP_001122101.1 |
C15orf56 - chromosome 15 open reading frame 56 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039905.2 | 2039 | UTR 3 | NP_001034994.1 |
PAK6 - p21 (RAC1) activated kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276717.1 | 2039 | Intron | NP_001263646.1 | |||
NM_001276718.1 | 2039 | Intron | NP_001263647.1 | |||
NM_020168.5 | 2039 | Intron | NP_064553.1 |