Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAACAAATACCCTGCTTCCTCACA[C/T]CCCCTTTTCTTCCTTGGAGCTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608110 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BUB1B-PAK6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BUB1B-PAK6 - BUB1B-PAK6 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128628.2 | Intron | NP_001122100.1 | ||||
NM_001128629.2 | Intron | NP_001122101.1 |
C15orf56 - chromosome 15 open reading frame 56 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAK6 - p21 (RAC1) activated kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276717.1 | Intron | NP_001263646.1 | ||||
NM_001276718.1 | Intron | NP_001263647.1 | ||||
NM_020168.5 | Intron | NP_064553.1 |