Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGAGTACATGGTAGCTGTATTT[A/G]TCAGACACTTAAAACTTGTAGCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf107 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C10orf107 - chromosome 10 open reading frame 107 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173554.2 | Intron | NP_775825.1 | ||||
XM_005269598.3 | Intron | XP_005269655.1 | ||||
XM_005269599.4 | Intron | XP_005269656.1 | ||||
XM_005269600.4 | Intron | XP_005269657.1 | ||||
XM_006717690.3 | Intron | XP_006717753.1 | ||||
XM_006717692.3 | Intron | XP_006717755.1 |