Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAAATCAGATAATCTGAGTGCTA[A/T]AATAGTTTGCCTCCTGGATTTATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611673 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRMT1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TRMT1L - tRNA methyltransferase 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202423.1 | 5428 | UTR 3 | NP_001189352.1 | |||
NM_030934.4 | 5428 | UTR 3 | NP_112196.3 | |||
XM_011510028.2 | 5428 | UTR 3 | XP_011508330.1 | |||
XM_017002419.1 | 5428 | Intron | XP_016857908.1 |
Set Membership: |
HapMap Validated |