Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTCACAGCCTGTGAAATTCCAA[C/T]CCATTTGTCTTCACCATTCCAACCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609424 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RC3H1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RC3H1 - ring finger and CCCH-type domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300850.1 | 10433 | UTR 3 | NP_001287779.1 | |||
NM_001300851.1 | 10433 | UTR 3 | NP_001287780.1 | |||
NM_001300852.1 | 10433 | UTR 3 | NP_001287781.1 | |||
NM_172071.3 | 10433 | UTR 3 | NP_742068.1 | |||
XM_005244921.3 | 10433 | UTR 3 | XP_005244978.1 | |||
XM_011509231.2 | 10433 | UTR 3 | XP_011507533.1 |