Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607369 MIM: 607370 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNK16 - potassium two pore domain channel subfamily K member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135105.1 | Intron | NP_001128577.1 | ||||
NM_001135106.1 | Intron | NP_001128578.1 | ||||
NM_001135107.1 | Intron | NP_001128579.1 | ||||
NM_032115.3 | Intron | NP_115491.1 | ||||
XM_011514935.2 | Intron | XP_011513237.1 | ||||
XM_017011346.1 | Intron | XP_016866835.1 |
KCNK17 - potassium two pore domain channel subfamily K member 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |