Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGGGATCTCCTCATGCCTAGCC[C/T]CATGCTTCTGCCTGAGGCATTTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610290 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALNT12 - polypeptide N-acetylgalactosaminyltransferase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024642.4 | 216 | Intron | NP_078918.3 | |||
XM_006717287.1 | 216 | Intron | XP_006717350.1 | |||
XM_011519018.2 | 216 | UTR 5 | XP_011517320.1 | |||
XM_011519020.1 | 216 | Intron | XP_011517322.1 | |||
XM_017015133.1 | 216 | Intron | XP_016870622.1 |