Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600006 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DCAF15 - DDB1 and CUL4 associated factor 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PODNL1 - podocan like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RFX1 - regulatory factor X1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002918.4 | 4489 | UTR 3 | NP_002909.4 | |||
XM_011528165.2 | 4489 | UTR 3 | XP_011526467.1 | |||
XM_011528166.2 | 4489 | UTR 3 | XP_011526468.1 | |||
XM_011528167.2 | 4489 | UTR 3 | XP_011526469.1 | |||
XM_011528168.2 | 4489 | UTR 3 | XP_011526470.1 | |||
XM_011528169.2 | 4489 | UTR 3 | XP_011526471.1 | |||
XM_011528170.2 | 4489 | UTR 3 | XP_011526472.1 |
Set Membership: |
HapMap |