Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604405 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIGLEC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SIGLEC6 - sialic acid binding Ig like lectin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011527533.2 | 3241 | UTR 3 | XP_011525835.1 | |||
XM_011527534.2 | 3241 | Intron | XP_011525836.1 | |||
XM_011527535.2 | 3241 | UTR 3 | XP_011525837.1 | |||
XM_011527536.2 | 3241 | UTR 3 | XP_011525838.1 | |||
XM_011527537.2 | 3241 | Intron | XP_011525839.1 | |||
XM_011527538.2 | 3241 | UTR 3 | XP_011525840.1 | |||
XM_011527539.2 | 3241 | Intron | XP_011525841.1 | |||
XM_017027511.1 | 3241 | Intron | XP_016883000.1 |
Set Membership: |
HapMap |