Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTAACTCATGAGTATTTGTGGC[G/T]CAAGGCTTGGAAGTGACCTTGGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614545 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMC10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EMC10 - ER membrane protein complex subunit 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175063.5 | Intron | NP_778233.4 | ||||
NM_206538.3 | Intron | NP_996261.1 | ||||
XM_006723163.3 | Intron | XP_006723226.1 | ||||
XM_011526818.2 | Intron | XP_011525120.1 |
FAM71E1 - family with sequence similarity 71 member E1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308429.1 | Intron | NP_001295358.1 | ||||
NM_138411.2 | Intron | NP_612420.1 | ||||
XM_011526405.2 | Intron | XP_011524707.2 | ||||
XM_011526406.2 | Intron | XP_011524708.2 |
Set Membership: |
HapMap Validated |