Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604996 MIM: 608303 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FXYD3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FXYD3 - FXYD domain containing ion transport regulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LGI4 - leucine rich repeat LGI family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139284.2 | Intron | NP_644813.1 | ||||
XM_011526595.1 | Intron | XP_011524897.1 | ||||
XM_017026428.1 | Intron | XP_016881917.1 | ||||
XM_017026429.1 | Intron | XP_016881918.1 | ||||
XM_017026430.1 | Intron | XP_016881919.1 |
MIR6887 - microRNA 6887 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |