Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGAAAACAGATCAGTTATTTAA[C/T]ATGTAAAAGAAATGTAGTTTTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601810 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DNA2 - DNA replication helicase/nuclease 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080449.2 | Intron | NP_001073918.2 | ||||
XM_006717680.3 | Intron | XP_006717743.1 | ||||
XM_011539417.1 | Intron | XP_011537719.1 | ||||
XM_017015799.1 | Intron | XP_016871288.1 |