Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTGCTTCAATATGTAAACAGTT[A/G]AAAGAAAGAAATTGTCAAAACATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601810 MIM: 139080 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNA2 - DNA replication helicase/nuclease 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A16 - solute carrier family 25 member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324312.1 | Intron | NP_001311241.1 | ||||
NM_001324313.1 | Intron | NP_001311242.1 | ||||
NM_001324314.1 | Intron | NP_001311243.1 | ||||
NM_001324315.1 | Intron | NP_001311244.1 | ||||
NM_001324317.1 | Intron | NP_001311246.1 | ||||
NM_152707.3 | Intron | NP_689920.1 | ||||
XM_011540211.2 | Intron | XP_011538513.1 | ||||
XM_017016695.1 | Intron | XP_016872184.1 |
Set Membership: |
HapMap |