Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATCAAATCTAGAGGACTCTCTT[A/G]AACTGGGAACCATAGCCACCAGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604515 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLNK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BLNK - B-cell linker | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114094.1 | Intron | NP_001107566.1 | ||||
NM_001258440.1 | Intron | NP_001245369.1 | ||||
NM_001258441.1 | Intron | NP_001245370.1 | ||||
NM_001258442.1 | Intron | NP_001245371.1 | ||||
NM_013314.3 | Intron | NP_037446.1 | ||||
XM_011539728.2 | Intron | XP_011538030.1 | ||||
XM_017016159.1 | Intron | XP_016871648.1 |
Set Membership: |
HapMap Validated |