Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTGAAAGTAAATATTTCTTACC[G/T]TGAATTTTGTTTATGCTATTTTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604515 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLNK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BLNK - B-cell linker | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114094.1 | Intron | NP_001107566.1 | ||||
NM_001258440.1 | Intron | NP_001245369.1 | ||||
NM_001258441.1 | Intron | NP_001245370.1 | ||||
NM_001258442.1 | Intron | NP_001245371.1 | ||||
NM_013314.3 | Intron | NP_037446.1 | ||||
XM_011539728.2 | Intron | XP_011538030.1 | ||||
XM_017016159.1 | Intron | XP_016871648.1 |
Set Membership: |
HapMap |