Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACAACCATTTTGGAAAACAATTT[A/G]GCATTTACTCCCAAAGCTGAACAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYPD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYPD4 - LY6/PLAUR domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291419.1 | 3322 | Intron | NP_001278348.1 | |||
NM_173506.6 | 3322 | Intron | NP_775777.3 | |||
XM_005258543.3 | 3322 | Intron | XP_005258600.1 | |||
XM_005278383.4 | 3322 | Intron | XP_005278440.1 | |||
XM_005278385.4 | 3322 | UTR 3 | XP_005278442.1 | |||
XM_005278387.4 | 3322 | Intron | XP_005278444.1 | |||
XM_005278388.3 | 3322 | Intron | XP_005278445.1 | |||
XM_006723035.2 | 3322 | Intron | XP_006723098.1 | |||
XM_011526483.2 | 3322 | Intron | XP_011524785.1 | |||
XM_011526484.2 | 3322 | Intron | XP_011524786.1 |