Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM213A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM213A - family with sequence similarity 213 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243778.1 | Intron | NP_001230707.1 | ||||
NM_001243779.1 | Intron | NP_001230708.1 | ||||
NM_001243780.1 | Intron | NP_001230709.1 | ||||
NM_001243781.1 | Intron | NP_001230710.1 | ||||
NM_001243782.1 | Intron | NP_001230711.1 | ||||
NM_032333.4 | Intron | NP_115709.3 | ||||
XM_011540265.2 | Intron | XP_011538567.1 | ||||
XM_011540266.2 | Intron | XP_011538568.1 | ||||
XM_017016775.1 | Intron | XP_016872264.1 |
Set Membership: |
HapMap |