Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604274 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLRA1P PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLRA1P - killer cell lectin like receptor A1, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAGOHB - mago homolog B, exon junction complex core component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300739.2 | 593 | UTR 3 | NP_001287668.1 | |||
NM_001319985.1 | 593 | UTR 3 | NP_001306914.1 | |||
NM_018048.4 | 593 | UTR 3 | NP_060518.1 | |||
XM_005253402.1 | 593 | Intron | XP_005253459.1 |