Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGTAGAGTTAAAGTTACAATCTC[C/T]TTCTACAACCTTATGAGAAATTTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600505 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSNK1A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
AMR
|
||||||||
EUR - Not Available |
CSNK1A1 - casein kinase 1 alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025105.2 | Intron | NP_001020276.1 | ||||
NM_001271741.1 | Intron | NP_001258670.1 | ||||
NM_001271742.1 | Intron | NP_001258671.1 | ||||
NM_001892.5 | Intron | NP_001883.4 |
Set Membership: |
HapMap |