Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTCCTTGGCAGCCAATTGAGAG[A/G]TGTAACATAATTGGCTCAGCCAATC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609804 MIM: 604203 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP52 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CFAP52 - cilia and flagella associated protein 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080556.1 | Intron | NP_001074025.1 | ||||
NM_145054.4 | Intron | NP_659491.4 | ||||
XM_017024227.1 | Intron | XP_016879716.1 |
STX8 - syntaxin 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |