Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606398 MIM: 609742 MIM: 605815 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATF5 - activating transcription factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193646.1 | 624 | Silent Mutation | CTA,TTA | L,L 83 | NP_001180575.1 | |
NM_001290746.1 | 624 | Silent Mutation | CTA,TTA | L,L 83 | NP_001277675.1 | |
NM_012068.5 | 624 | Silent Mutation | CTA,TTA | L,L 83 | NP_036200.2 | |
XM_011526629.2 | 624 | Silent Mutation | CTA,TTA | L,L 83 | XP_011524931.1 |
IL4I1 - interleukin 4 induced 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4751 - microRNA 4751 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUP62 - nucleoporin 62 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |