Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616702 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF589 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF589 - zinc finger protein 589 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016089.2 | 1436 | Intron | NP_057173.2 | |||
XM_017006575.1 | 1436 | Intron | XP_016862064.1 | |||
XM_017006576.1 | 1436 | Silent Mutation | TGC,TGT | C,C 26 | XP_016862065.1 | |
XM_017006577.1 | 1436 | Silent Mutation | TGC,TGT | C,C 26 | XP_016862066.1 | |
XM_017006578.1 | 1436 | Silent Mutation | TGC,TGT | C,C 26 | XP_016862067.1 | |
XM_017006579.1 | 1436 | Silent Mutation | TGC,TGT | C,C 26 | XP_016862068.1 |
Set Membership: |
HapMap |