Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600429 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCNT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GCNT2 - glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001491.2 | Intron | NP_001482.1 | ||||
NM_145649.4 | Intron | NP_663624.1 | ||||
NM_145655.3 | Intron | NP_663630.2 | ||||
XM_005248997.3 | Intron | XP_005249054.1 | ||||
XM_005248999.2 | Intron | XP_005249056.1 | ||||
XM_006715052.3 | Intron | XP_006715115.1 | ||||
XM_011514466.2 | Intron | XP_011512768.1 | ||||
XM_011514468.2 | Intron | XP_011512770.1 | ||||
XM_017010732.1 | Intron | XP_016866221.1 |
Set Membership: |
Validated |