Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTATGTTTTACCAGTGTGAAAT[A/T]TTATGTCTAATTAAAAAAAAAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615929 MIM: 610428 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD17 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKRD17 - ankyrin repeat domain 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286771.2 | 8672 | UTR 3 | NP_001273700.1 | |||
NM_015574.1 | 8672 | UTR 3 | NP_056389.1 | |||
NM_032217.4 | 8672 | UTR 3 | NP_115593.3 | |||
NM_198889.2 | 8672 | UTR 3 | NP_942592.1 | |||
XM_005265671.3 | 8672 | Intron | XP_005265728.1 | |||
XM_005265672.3 | 8672 | Intron | XP_005265729.1 | |||
XM_005265673.3 | 8672 | Intron | XP_005265730.1 | |||
XM_017008011.1 | 8672 | Intron | XP_016863500.1 | |||
XM_017008012.1 | 8672 | Intron | XP_016863501.1 | |||
XM_017008013.1 | 8672 | Intron | XP_016863502.1 | |||
XM_017008014.1 | 8672 | Intron | XP_016863503.1 | |||
XM_017008015.1 | 8672 | Intron | XP_016863504.1 | |||
XM_017008016.1 | 8672 | Intron | XP_016863505.1 | |||
XM_017008017.1 | 8672 | Intron | XP_016863506.1 |
COX18 - COX18, cytochrome c oxidase assembly factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |