Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610428 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX18 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COX18 - COX18, cytochrome c oxidase assembly factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297732.1 | 4220 | UTR 3 | NP_001284661.1 | |||
NM_001297733.1 | 4220 | UTR 3 | NP_001284662.1 | |||
NM_001300729.1 | 4220 | UTR 3 | NP_001287658.1 | |||
NM_173827.3 | 4220 | UTR 3 | NP_776188.1 | |||
XM_005265680.4 | 4220 | Intron | XP_005265737.1 | |||
XM_011531878.2 | 4220 | Intron | XP_011530180.1 | |||
XM_017008045.1 | 4220 | Intron | XP_016863534.1 |
Set Membership: |
HapMap |