Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATGGTTCATCTATGGTGAACTG[C/T]ACTCAACACTGGATGGATGATAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD18 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ABHD18 - abhydrolase domain containing 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319305.1 | Intron | NP_001306234.1 | ||||
NM_001319306.1 | Intron | NP_001306235.1 | ||||
NM_001319307.1 | Intron | NP_001306236.1 | ||||
XM_005263238.4 | Intron | XP_005263295.1 | ||||
XM_006714318.3 | Intron | XP_006714381.1 | ||||
XM_006714320.3 | Intron | XP_006714383.1 | ||||
XM_006714322.3 | Intron | XP_006714385.1 | ||||
XM_011532272.2 | Intron | XP_011530574.1 | ||||
XM_011532273.2 | Intron | XP_011530575.1 | ||||
XM_011532274.2 | Intron | XP_011530576.1 | ||||
XM_011532276.1 | Intron | XP_011530578.1 | ||||
XM_011532278.2 | Intron | XP_011530580.1 | ||||
XM_011532279.2 | Intron | XP_011530581.1 | ||||
XM_017008631.1 | Intron | XP_016864120.1 | ||||
XM_017008632.1 | Intron | XP_016864121.1 |