Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTGTCAATTAGTAACTCTTTAC[A/T]ATATAATTAAAAATTTGAAGAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612889 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLJ27354 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FLJ27354 - uncharacterized LOC400761 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC8C - leucine rich repeat containing 8 family member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032270.4 | Intron | NP_115646.2 | ||||
XM_006710960.3 | Intron | XP_006711023.1 | ||||
XM_011542282.2 | Intron | XP_011540584.1 | ||||
XM_011542283.2 | Intron | XP_011540585.1 | ||||
XM_017002503.1 | Intron | XP_016857992.1 | ||||
XM_017002504.1 | Intron | XP_016857993.1 |
Set Membership: |
Validated |