Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGGCTCTTCTTCAGAAACATTTG[C/T]CTTTTGATCATTCTAATGGTAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607892 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DSG1-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DSG1-AS1 - DSG1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DSG4 - desmoglein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134453.2 | 165 | Silent Mutation | TGC,TGT | C,C 10 | NP_001127925.1 | |
NM_177986.4 | 165 | Silent Mutation | TGC,TGT | C,C 10 | NP_817123.1 | |
XM_011525827.1 | 165 | Intron | XP_011524129.1 |
Set Membership: |
HapMap |