Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATTTGCTTTACATTCGGTCGGTC[C/T]GTCTTAAATAGGGATTTCCCCTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612814 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATA18 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPATA18 - spermatogenesis associated 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297608.1 | Intron | NP_001284537.1 | ||||
NM_145263.3 | Intron | NP_660306.1 | ||||
XM_005265716.1 | Intron | XP_005265773.1 | ||||
XM_005265717.2 | Intron | XP_005265774.1 | ||||
XM_005265718.1 | Intron | XP_005265775.1 | ||||
XM_011534369.1 | Intron | XP_011532671.1 | ||||
XM_011534370.1 | Intron | XP_011532672.1 | ||||
XM_011534372.1 | Intron | XP_011532674.1 | ||||
XM_017007731.1 | Intron | XP_016863220.1 |