Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCATGAAGACTTCCTGATCAGCCC[A/G]TCCCAAGAGACTATGTACAGTCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
60 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153420 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD58 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD58 - CD58 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144822.1 | 15843 | Intron | NP_001138294.1 | |||
NM_001779.2 | 15843 | Intron | NP_001770.1 | |||
XM_017002869.1 | 15843 | UTR 3 | XP_016858358.1 |
LOC105378922 - uncharacterized LOC105378922 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |