Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGCTAAATACAAGTCACAGGCT[C/T]AGTAAGTTAAGACAACATACAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
73 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608917 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATPAF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATPAF1 - ATP synthase mitochondrial F1 complex assembly factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EFCAB14 - EF-hand calcium binding domain 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014774.2 | 4880 | UTR 3 | NP_055589.1 |
EFCAB14-AS1 - EFCAB14 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX38 - testis expressed 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |