Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTCCAGGGGGAATTTTGATGTA[C/T]CACAAAAACCAGCATCCAGTTGATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609543 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00293 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LINC00293 - long intergenic non-protein coding RNA 293 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |