Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAAAAAAAAAAAAAAAATGAATC[A/G]ATTATGTTACTGATATTCTTCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114815 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CA8 - carbonic anhydrase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321837.1 | Intron | NP_001308766.1 | ||||
NM_001321838.1 | Intron | NP_001308767.1 | ||||
NM_001321839.1 | Intron | NP_001308768.1 | ||||
NM_004056.5 | Intron | NP_004047.3 | ||||
XM_011517587.2 | Intron | XP_011515889.1 | ||||
XM_011517588.2 | Intron | XP_011515890.1 | ||||
XM_017013818.1 | Intron | XP_016869307.1 |