Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610010 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARSJ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARSJ - arylsulfatase family member J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024590.3 | 4228 | UTR 3 | NP_078866.3 | |||
XM_005263212.4 | 4228 | UTR 3 | XP_005263269.1 | |||
XM_011532238.2 | 4228 | UTR 3 | XP_011530540.1 | |||
XM_011532239.2 | 4228 | UTR 3 | XP_011530541.1 | |||
XM_017008592.1 | 4228 | UTR 3 | XP_016864081.1 | |||
XM_017008593.1 | 4228 | UTR 3 | XP_016864082.1 | |||
XM_017008594.1 | 4228 | UTR 3 | XP_016864083.1 | |||
XM_017008595.1 | 4228 | UTR 3 | XP_016864084.1 | |||
XM_017008596.1 | 4228 | UTR 3 | XP_016864085.1 | |||
XM_017008597.1 | 4228 | UTR 3 | XP_016864086.1 | |||
XM_017008598.1 | 4228 | UTR 3 | XP_016864087.1 | |||
XM_017008599.1 | 4228 | UTR 3 | XP_016864088.1 | |||
XM_017008600.1 | 4228 | UTR 3 | XP_016864089.1 |