Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609295 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEMA6D PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEMA6D - semaphorin 6D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198999.1 | Intron | NP_001185928.1 | ||||
NM_020858.1 | Intron | NP_065909.1 | ||||
NM_024966.2 | Intron | NP_079242.2 | ||||
NM_153616.1 | Intron | NP_705869.1 | ||||
NM_153617.1 | Intron | NP_705870.1 | ||||
NM_153618.1 | Intron | NP_705871.1 | ||||
NM_153619.1 | Intron | NP_705872.1 | ||||
XM_005254685.3 | Intron | XP_005254742.1 | ||||
XM_005254686.2 | Intron | XP_005254743.1 | ||||
XM_005254687.2 | Intron | XP_005254744.1 | ||||
XM_005254689.3 | Intron | XP_005254746.1 | ||||
XM_011522075.2 | Intron | XP_011520377.1 | ||||
XM_011522076.2 | Intron | XP_011520378.1 | ||||
XM_011522077.2 | Intron | XP_011520379.1 | ||||
XM_011522078.2 | Intron | XP_011520380.1 | ||||
XM_011522079.2 | Intron | XP_011520381.1 | ||||
XM_011522080.2 | Intron | XP_011520382.1 | ||||
XM_011522081.2 | Intron | XP_011520383.1 | ||||
XM_017022617.1 | Intron | XP_016878106.1 | ||||
XM_017022618.1 | Intron | XP_016878107.1 | ||||
XM_017022619.1 | Intron | XP_016878108.1 | ||||
XM_017022620.1 | Intron | XP_016878109.1 | ||||
XM_017022621.1 | Intron | XP_016878110.1 | ||||
XM_017022622.1 | Intron | XP_016878111.1 |
Set Membership: |
HapMap |