Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGAATTTATCTGAATATTTTGCT[A/T]GTCTGAATTGCTAATTTGTCAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603331 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYNC1I2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DYNC1I2 - dynein cytoplasmic 1 intermediate chain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271785.1 | Intron | NP_001258714.1 | ||||
NM_001271786.1 | Intron | NP_001258715.1 | ||||
NM_001271787.1 | Intron | NP_001258716.1 | ||||
NM_001271788.1 | Intron | NP_001258717.1 | ||||
NM_001271789.1 | Intron | NP_001258718.1 | ||||
NM_001271790.1 | Intron | NP_001258719.1 | ||||
NM_001320882.1 | Intron | NP_001307811.1 | ||||
NM_001320883.1 | Intron | NP_001307812.1 | ||||
NM_001320884.1 | Intron | NP_001307813.1 | ||||
NM_001378.2 | Intron | NP_001369.1 | ||||
XM_006712347.3 | Intron | XP_006712410.1 | ||||
XM_017003525.1 | Intron | XP_016859014.1 |
Set Membership: |
HapMap |