Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151525 MIM: 615354 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD53 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CD53 - CD53 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000560.3 | 2607 | Intron | NP_000551.1 | |||
NM_001040033.1 | 2607 | Intron | NP_001035122.1 | |||
NM_001320638.1 | 2607 | Intron | NP_001307567.1 |
LRIF1 - ligand dependent nuclear receptor interacting factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006945.1 | 2607 | Intron | NP_001006946.1 | |||
NM_018372.3 | 2607 | Intron | NP_060842.3 | |||
XM_005271029.3 | 2607 | Intron | XP_005271086.1 | |||
XM_011541768.1 | 2607 | Intron | XP_011540070.1 | |||
XM_017001768.1 | 2607 | Intron | XP_016857257.1 | |||
XM_017001769.1 | 2607 | UTR 3 | XP_016857258.1 |