Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGACCATTTTATTAGTTCATGAC[A/G]CAAAGATTAGGAGGCGGTCCATACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608692 MIM: 613465 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLZF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BLZF1 - basic leucine zipper nuclear factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320972.1 | Intron | NP_001307901.1 | ||||
NM_001320973.1 | Intron | NP_001307902.1 | ||||
NM_003666.3 | Intron | NP_003657.1 | ||||
XM_005245561.4 | Intron | XP_005245618.1 | ||||
XM_011510092.2 | Intron | XP_011508394.1 | ||||
XM_017002645.1 | Intron | XP_016858134.1 |
NME7 - NME/NM23 family member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |