Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608674 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH3D19 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SH3D19 - SH3 domain containing 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009555.3 | 5123 | UTR 3 | NP_001009555.3 | |||
NM_001128923.1 | 5123 | UTR 3 | NP_001122395.1 | |||
NM_001128924.1 | 5123 | UTR 3 | NP_001122396.1 | |||
NM_001243349.1 | 5123 | UTR 3 | NP_001230278.1 | |||
XM_005262767.2 | 5123 | UTR 3 | XP_005262824.1 | |||
XM_011531646.1 | 5123 | UTR 3 | XP_011529948.1 | |||
XM_011531647.1 | 5123 | UTR 3 | XP_011529949.1 | |||
XM_011531648.1 | 5123 | UTR 3 | XP_011529950.1 | |||
XM_011531649.1 | 5123 | UTR 3 | XP_011529951.1 | |||
XM_017007778.1 | 5123 | UTR 3 | XP_016863267.1 | |||
XM_017007779.1 | 5123 | UTR 3 | XP_016863268.1 | |||
XM_017007780.1 | 5123 | UTR 3 | XP_016863269.1 | |||
XM_017007781.1 | 5123 | UTR 3 | XP_016863270.1 | |||
XM_017007782.1 | 5123 | UTR 3 | XP_016863271.1 | |||
XM_017007783.1 | 5123 | UTR 3 | XP_016863272.1 |