Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609230 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NUDT5 - nudix hydrolase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321647.1 | 2470 | UTR 3 | NP_001308576.1 | |||
NM_001321648.1 | 2470 | UTR 3 | NP_001308577.1 | |||
NM_014142.3 | 2470 | UTR 3 | NP_054861.2 |
SEC61A2 - Sec61 translocon alpha 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142627.2 | 2470 | Intron | NP_001136099.1 | |||
NM_001142628.1 | 2470 | Intron | NP_001136100.1 | |||
NM_018144.3 | 2470 | Intron | NP_060614.2 |
Set Membership: |
HapMap Validated |