Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTATTGAGATTATAATTCAACT[A/G]TCATGATTTATTCATTTAAAGTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615843 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEGS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DEGS1 - delta 4-desaturase, sphingolipid 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321541.1 | Intron | NP_001308470.1 | ||||
NM_001321542.1 | Intron | NP_001308471.1 | ||||
NM_003676.3 | Intron | NP_003667.1 | ||||
XM_017002648.1 | Intron | XP_016858137.1 |
Set Membership: |
HapMap |