Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605511 MIM: 605736 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMPRSS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TMPRSS3 - transmembrane protease, serine 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBASH3A - ubiquitin associated and SH3 domain containing A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001895.2 | 94 | Missense Mutation | AGC,GGC | S,G 18 | NP_001001895.1 | |
NM_001243467.1 | 94 | Missense Mutation | AGC,GGC | S,G 18 | NP_001230396.1 | |
NM_018961.3 | 94 | Missense Mutation | AGC,GGC | S,G 18 | NP_061834.1 | |
XM_006724013.3 | 94 | UTR 5 | XP_006724076.1 | |||
XM_011529605.2 | 94 | Missense Mutation | AGC,GGC | S,G 18 | XP_011527907.1 | |
XM_011529606.2 | 94 | UTR 5 | XP_011527908.1 | |||
XM_011529607.2 | 94 | UTR 5 | XP_011527909.1 | |||
XM_011529609.1 | 94 | Missense Mutation | AGC,GGC | S,G 18 | XP_011527911.1 | |
XM_011529610.2 | 94 | Intron | XP_011527912.1 |