Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605924 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
WDR4 - WD repeat domain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260474.1 | 1802 | UTR 3 | NP_001247403.1 | |||
NM_001260475.1 | 1802 | UTR 3 | NP_001247404.1 | |||
NM_001260476.1 | 1802 | UTR 3 | NP_001247405.1 | |||
NM_001260477.1 | 1802 | UTR 3 | NP_001247406.1 | |||
NM_018669.5 | 1802 | UTR 3 | NP_061139.2 | |||
NM_033661.4 | 1802 | Intron | NP_387510.1 | |||
XM_011529433.1 | 1802 | UTR 3 | XP_011527735.1 | |||
XM_017028262.1 | 1802 | UTR 3 | XP_016883751.1 | |||
XM_017028263.1 | 1802 | UTR 3 | XP_016883752.1 | |||
XM_017028264.1 | 1802 | Intron | XP_016883753.1 |
Set Membership: |
HapMap Validated |