Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120930 MIM: 609072 MIM: 612905 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8G PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C8G - complement component 8, gamma polypeptide | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXW5 - F-box and WD repeat domain containing 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LCN12 - lipocalin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178536.3 | Intron | NP_848631.2 | ||||
XM_006717065.3 | Intron | XP_006717128.1 | ||||
XM_011518560.2 | Intron | XP_011516862.1 | ||||
XM_011518561.2 | Intron | XP_011516863.2 | ||||
XM_011518562.2 | Intron | XP_011516864.1 | ||||
XM_017014631.1 | Intron | XP_016870120.1 | ||||
XM_017014632.1 | Intron | XP_016870121.1 |
Set Membership: |
HapMap |