Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTCAGCCTCCACCTGGTTCCTG[C/T]AGGCTGGGGGTGACTTCTTCCTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604184 MIM: 616810 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGDCC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGDCC3 - immunoglobulin superfamily DCC subclass member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IGDCC4 - immunoglobulin superfamily DCC subclass member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020962.2 | 3391 | Missense Mutation | TAC,TGC | Y,C 1125 | NP_066013.1 | |
XM_011521845.2 | 3391 | Missense Mutation | TAC,TGC | Y,C 1138 | XP_011520147.1 | |
XM_011521846.2 | 3391 | Missense Mutation | TAC,TGC | Y,C 1126 | XP_011520148.1 | |
XM_011521847.1 | 3391 | Missense Mutation | TAC,TGC | Y,C 864 | XP_011520149.1 | |
XM_017022448.1 | 3391 | Missense Mutation | TAC,TGC | Y,C 1139 | XP_016877937.1 | |
XM_017022449.1 | 3391 | Intron | XP_016877938.1 |