Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186854 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GRIP2 - glutamate receptor interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080423.3 | 6198 | UTR 3 | NP_001073892.3 | |||
XM_011534141.2 | 6198 | Intron | XP_011532443.1 | |||
XM_011534142.2 | 6198 | Intron | XP_011532444.1 | |||
XM_011534143.2 | 6198 | Intron | XP_011532445.1 | |||
XM_011534145.2 | 6198 | Intron | XP_011532447.1 | |||
XM_011534146.2 | 6198 | Intron | XP_011532448.1 |
SLC6A6 - solute carrier family 6 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134367.3 | 6198 | UTR 3 | NP_001127839.2 | |||
NM_001134368.3 | 6198 | Intron | NP_001127840.1 | |||
NM_003043.5 | 6198 | UTR 3 | NP_003034.2 | |||
XM_006713307.2 | 6198 | UTR 3 | XP_006713370.1 | |||
XM_011534030.2 | 6198 | UTR 3 | XP_011532332.1 | |||
XM_011534031.2 | 6198 | UTR 3 | XP_011532333.1 |
Set Membership: |
HapMap Validated |