Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGAATTTCCAGGTCAGGGATTT[C/T]ACATTACATCTGCTTAGGTGTGGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616713 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIPOX PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PIPOX - pipecolic acid and sarcosine oxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016518.2 | Intron | NP_057602.2 | ||||
XM_017024722.1 | Intron | XP_016880211.1 |
Set Membership: |
HapMap Validated |