Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAAAACATGCCCAGAAGTTTCC[A/T]GCTTAGATGATGAAGTTCAATGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612344 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF385B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF385B - zinc finger protein 385B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113397.1 | Intron | NP_001106868.1 | ||||
NM_001113398.1 | Intron | NP_001106869.1 | ||||
NM_001282725.1 | Intron | NP_001269654.1 | ||||
NM_152520.4 | Intron | NP_689733.3 | ||||
XM_011510713.2 | Intron | XP_011509015.1 | ||||
XM_011510714.2 | Intron | XP_011509016.1 | ||||
XM_011510715.2 | Intron | XP_011509017.1 | ||||
XM_011510716.2 | Intron | XP_011509018.1 | ||||
XM_011510717.2 | Intron | XP_011509019.1 | ||||
XM_011510719.2 | Intron | XP_011509021.1 | ||||
XM_011510720.2 | Intron | XP_011509022.1 | ||||
XM_011510721.2 | Intron | XP_011509023.1 | ||||
XM_011510723.2 | Intron | XP_011509025.1 | ||||
XM_017003435.1 | Intron | XP_016858924.1 | ||||
XM_017003436.1 | Intron | XP_016858925.1 | ||||
XM_017003437.1 | Intron | XP_016858926.1 | ||||
XM_017003438.1 | Intron | XP_016858927.1 | ||||
XM_017003439.1 | Intron | XP_016858928.1 | ||||
XM_017003440.1 | Intron | XP_016858929.1 | ||||
XM_017003441.1 | Intron | XP_016858930.1 |
Set Membership: |
HapMap |