Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGATGAATAAAAATACTGAACTG[G/T]GCCTTTAAAAAAGTACAGAAAATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606409 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITCH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ITCH - itchy E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257137.2 | Intron | NP_001244066.1 | ||||
NM_001257138.2 | Intron | NP_001244067.1 | ||||
NM_001324197.1 | Intron | NP_001311126.1 | ||||
NM_001324198.1 | Intron | NP_001311127.1 | ||||
NM_031483.6 | Intron | NP_113671.3 | ||||
XM_005260570.1 | Intron | XP_005260627.1 | ||||
XM_011529079.1 | Intron | XP_011527381.1 | ||||
XM_017028089.1 | Intron | XP_016883578.1 | ||||
XM_017028090.1 | Intron | XP_016883579.1 | ||||
XM_017028091.1 | Intron | XP_016883580.1 | ||||
XM_017028092.1 | Intron | XP_016883581.1 |
Set Membership: |
Validated |