Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCAGCATCAGGAGACTGGCCTC[A/C]AGGCTGGAGCCACCCCCACACCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613535 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0319L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIAA0319L - KIAA0319 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024874.4 | Intron | NP_079150.3 | ||||
XM_006710907.1 | Intron | XP_006710970.1 | ||||
XM_006710909.1 | Intron | XP_006710972.1 | ||||
XM_006710910.1 | Intron | XP_006710973.1 | ||||
XM_011542179.2 | Intron | XP_011540481.1 | ||||
XM_011542180.1 | Intron | XP_011540482.1 | ||||
XM_017002366.1 | Intron | XP_016857855.1 | ||||
XM_017002367.1 | Intron | XP_016857856.1 | ||||
XM_017002368.1 | Intron | XP_016857857.1 | ||||
XM_017002369.1 | Intron | XP_016857858.1 | ||||
XM_017002370.1 | Intron | XP_016857859.1 | ||||
XM_017002371.1 | Intron | XP_016857860.1 | ||||
XM_017002372.1 | Intron | XP_016857861.1 | ||||
XM_017002373.1 | Intron | XP_016857862.1 | ||||
XM_017002374.1 | Intron | XP_016857863.1 | ||||
XM_017002375.1 | Intron | XP_016857864.1 | ||||
XM_017002376.1 | Intron | XP_016857865.1 | ||||
XM_017002377.1 | Intron | XP_016857866.1 | ||||
XM_017002378.1 | Intron | XP_016857867.1 |